missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDNF Antibody (35928.11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
215.00 EUR
Specifications
| Antigen | BDNF |
|---|---|
| Clone | 35928.11 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1 - 2 μg/mL, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 8 - 25 μg/mL |
| Applications | Western Blot |
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| BDNF | |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cytokine Research, Immunology, Neuroscience | |
| abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
| BDNF | |
| IgG1 | |
| Protein A purified | |
| Monoclonal Anti-Human BDNF will neutralize the biological activity of rhBDNF. This antibody shows < 2% cross-reactivity with rhbeta-NGF, rhNT-3 and rhNT-4 based on ELISA and immunoblotting. |
| 35928.11 | |
| Western Blot 1 - 2 μg/mL, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 8 - 25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 627 | |
| Recombinant human BDNF (rhBDNF) expressed in Sf 21 insect cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title