missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDNF Antibody (35928.11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB120-10505-25ug
This item is not returnable.
View return policy
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| BDNF | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1 - 2 μg/mL, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 8 - 25 μg/mL | |
| BDNF | |
| Recombinant human BDNF (rhBDNF) expressed in Sf 21 insect cells. | |
| 25 μg | |
| Cytokine Research, Immunology, Neuroscience | |
| 627 | |
| Human | |
| Purified |
| Western Blot | |
| 35928.11 | |
| Unconjugated | |
| abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Monoclonal Anti-Human BDNF will neutralize the biological activity of rhBDNF. This antibody shows < 2% cross-reactivity with rhbeta-NGF, rhNT-3 and rhNT-4 based on ELISA and immunoblotting. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction