Learn More
BD Violet Proliferation Dye 450
Product Code. 15180577
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15180577 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15180577 Supplier BD Supplier No. 562158
 Restricted Item
Estimated Shipment: 12-11-2026
For monitoring of cell divisions completed by single cells, intracellular staining
Flow Cytometry, Intracellular staining
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg
Content And Storage -20°C
For Use With (Application) Intracellular Staining, Monitoring Cell Divisions