Learn More
BD Retrievagen Antigen Retrieval Systems
Product Code. 15811807 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
125 mL
250 mL
Reagent Type:
Retrievagen A
Retrievagen B
Unit Size:
1 L
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Reagent Type Unit Size
15811807 125 mL Retrievagen A 1 L
15806308 250 mL Retrievagen B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15811807 Supplier BD Supplier No. 550524
 Restricted Item
Estimated Shipment: 01-12-2026
Recommended for zinc-fixed and formalin-fixed paraffin embedded tissue sections that show suboptimal or no reactivity with antibodies for immunohistochemical staining
10% neutral buffered formalin fixes the tissue by cross-linking proteins that may cause a conformational change or “masking” of the antigenic epitope leading to reduced or absent antibody reactivity. The BD Retrievagen Antigen Retrieval Systems are designed to unmask the epitope and cause the enhancement of antibody staining.
  • Retrievagen A (pH 6.0) is provided as two separate solutions that need to be mixed
  • Retrievagen B (pH 9.5) is provided as a ready-to-use solution
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 25mL Retrievagen A Solution 1 and 100mL Retrievagen A Solution 2
Dilution Take 18 mL of BD Retrievagen A Solution 1 and 82 mL of BD Retrievagen Solution 2 and bring up the volume to 1 L with distilled Water
Product Type Retrieval Reagent
Quantity 125 mL
For Use With (Application) Immunohistochemical staining
pH 6
Regulatory Status RUO
Reagent Type Retrievagen A