Learn More
BD Phospholipid-binding Protein, APC Annexin V
Product Code. 15856278
Change view
Click to view available options
Quantity:
100 Tests
200 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15856278 100 Tests Each
15866278 200 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15856278 Supplier BD Supplier No. 550474
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
Has high affinity for PS, and binds to cells with exposed PS
  • Conjugated to APC
  • Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 Tests
Sample Volume 5 μL
Storage Buffer Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Conjugate APC (Allophycocyanin)
Form Liquid