Learn More
BD P19 Cell Lysate
Product Code. 15806009
Click to view available options
Quantity:
500 μg
Unit Size:
Each
Product Code. 15806009
Brand: BD 611673
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Mouse Embryonal Carcinoma
Convenient to use and provided in denatured format allowing for immediate loading onto SDS-PAGE gel
  • Useful control for western blotting application
  • Rich in endogenously expressed proteins
  • Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
TRUSTED_SUSTAINABILITY
Specifications
Tissue Embryonal Carcinoma
Quantity 500 μg
Storage Buffer 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Host Species Mouse
Concentration 1 mg/mL
Content And Storage -20°C