missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD OptEIA™ Reagent Set B
Click to view available options
Quantity:
1 Set
Unit Size:
Each
Description
Includes
Wash Concentrate 20X, substrate reagent A, substrate reagent B, stop solution, assay diluent, coating buffer
Enzyme-linked immunosorbent assay
Specifications
Specifications
| Product Type | OptEIA™ reagent set B |
| Quantity | 1 Set |
| For Use With (Application) | ELISA |
| Content And Storage | 2°C to 8°C |
| Kit Contents | 250mL Coating buffer pH 9.5, 1L Assay diluent, 1L Wash concentrate 20X, 125mL Substrate reagent A, 125mL Reagent B, 125mL Stop solution |
| Sufficient For | 20 96-well Plates |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction