Learn More
BD Macro-Vue™ RPR Cards and Components
Product Code. 17352421
Change view
Click to view available options
Quantity:
10 Pack
Type:
RPR Control Cards
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Type Unit Size
17352421 10 Pack RPR Control Cards Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17352421 Supplier BD Supplier No. 276709
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For large-scale testing in public health and clinical laboratories
RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.
  • CLIA Complexity: Moderate
  • Nontreponemal test
  • For the serological detection of syphilis as described in A Manual of Tests for Syphilis†
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Reagin (Syphilis)
Type RPR Control Cards
For Use With (Application) For large-scale testing in public health and clinical laboratories
Includes BD Macro-Vue RPR control cards, 18-mm circle
Quantity 10 Pack