Learn More
BD BV480 Mouse Anti-Human CD3
Product Code. 15814809
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
Product Code. 15814809
Brand: BD 566166
CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. BD Horizon™ BV480 Mouse Anti-Human CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε
  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY
Specifications
Product Type BV480 Mouse Anti-Human CD3
Quantity 25 Tests
For Use With (Application) Flow Cytometry
Cross Reactivity QC Testing: Human
Form BV480
Sample Volume 5 μL
Content And Storage Store undiluted at 4°C
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide