Learn More
BD GasPak™ EZ Gas Generating Systems Incubation Containers
Product Code. 10118924
Change view
Click to view available options
Holds:
Petri Dishes (15 to 18)
Petri Dishes (30 to 33)
Size:
Large
Standard
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Size Unit Size
10118924 Petri Dishes (15 to 18) Standard Each
11737194 Petri Dishes (30 to 33) Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10118924 Supplier BD Supplier No. 260671
 Restricted Item
Estimated Shipment: 15-12-2026
Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
  • Non-breakable, chemical resistant containers
  • Removable Petri dish racks are available (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Type EZ Incubation Container
Closure Type Easy close latches
Content And Storage 2°C to 25°C
Disposable Reusable
Format Complete Box
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Holds Petri Dishes (15 to 18)
Product Line GasPak
Quantity Each
Size Standard