Learn More
BD Disposable Syringes with Luer-Lok™ Tips
Product Code. 10630694
Change view
Click to view available options
Volume (Metric):
1 mL
Unit Size:
Pack of 100
Pack of 800
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10630694 1 mL Pack of 100
15880305 1 mL Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10630694 Supplier BD Supplier No. 309628
  • ONLINE EXCLUSIVE PRICE
    506.68 EUR
    240.01 EUR / Pack of 100
    Save 266.67 EUR
    52% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
  • Sterile syringe for single use
  • Without needle
  • Clear barrel (polycarbonate or polypropylene) with bold scale markings
  • Tapered plunger rod for ease of aspiration
  • Positive plunger rod stop
  • Added BD Luer-Lok™ thread for increased secure connection
  • DNR (Latex) free
  • Shelf life: 1825 days
TRUSTED_SUSTAINABILITY
Specifications
Material Polycarbonate
Product Type Syringe
Disposable Single-use
Needle Point Number No Needle Included
Volume (Metric) 1 mL
Connection Type Luer Lock
Graduated Yes