Learn More
BD Difco™ Dehydrated Culture Media: Pseudomonas Isolation Agar
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Pseudomonas Isolation Agar |
| Product Type | Agar |
| Formulation | Peptone 20g/L, Magnesium Chloride 1.4g/L, Potassium Sulfate 10g/L, Irgasan™ 25mg/L, Agar 13.6g/L |
| For Use With (Application) | Used with Glycerol for selective isolation and differentiation of P. aeruginosa based on pigment formation |
| Packaging Type | Bottle |
| Quantity | 500 g |