Learn More
BD Difco™ Dehydrated Culture Media: Granulated Agar
Product Code. 11718233
Change view
Click to view available options
Quantity:
500 g
2000 g
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11718233 2000 g 2kg
10006334 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11718233 Supplier BD Supplier No. 214510
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Used as a solidifying agent for culture media. BD Difco™ Dehydrated Culture Media: Granulated Agar is carefully monitored for cultural response, solubility and gelation temperature.
Applications:
  • Solidifying agent for culture media
  • Granulated
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO/DIS, 11014
Concentration 1.5%
Additive Agar
Form Dehydrated Granule
For Use With (Application) General bacteriological purposes
Packaging Type Container
Quantity 2000 g