missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) RAT IL-1α; Flex Set, Bead A4
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
RAT IL-1α; Standard; RAT IL-1α; PE Detection Reagent; RAT IL-1α; Capture Bead A4
The BD CBA RAT IL-1α Flex Set is a bead-based immunoassay capable of measuring rat interleukin-1α (IL-1α) in serum and cell culture supernatant samples.
- Reactivity: QC Tested: Rat
- Bead Position: A4
- Limit of detection: 4.0pg/mL
With BD CBA Mouse/Rat Soluble Protein Master Buffer Kit (Cat. No. 558266, 100 tests, or 558267, 500 tests), a flow cytometer, and the FCAP Array™ software
Specifications
Specifications
| Antigen | IL-1alpha |
| Cross Reactivity | Rat |
| Includes | 1 Vial capture bead, 1 vial detection reagent and 2 vials standard |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Rat IL-1alpha flex set (bead A4) |
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction