Learn More
BD Cytometric Bead Array (CBA) Mouse RANTES; Flex Set, Bead D8
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Mouse RANTES; Standard; Mouse RANTES; PE Detection Reagent; Mouse RANTES; Capture Bead D8
Mouse reactivity is determined by testing samples with the BD CBA Mouse RANTES Flex Set.
Specifications
Specifications
| Antigen | RANTES |
| Cross Reactivity | Mouse |
| Includes | Mouse RANTES, standard, Mouse RANTES, PE detection reagent, Mouse RANTES, capture bead D8 |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse RANTES flex set (bead D8) |