missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse CD62E (E-Selectin); Flex Set, Bead E6
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
- Mouse soluble CD62E (E-Selectin) Standard
- Mouse CD62E (E-Selectin) PE Detection Reagent
- Mouse CD62E (E-Selectin) Capture Bead E6
Cotains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. Standard component is lyophilized and should be transferred to 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL assay Diluent, the standard has a protein concentration of 10,000pg/mL.
- Mouse reactivity was determined by testing samples with this Flex set
- Must be used in conjunction with CBA Mouse/Rat Soluble Protein Master Buffer Kit, flow cytometer and FCAP Array Software
Specifications
Specifications
| Antigen | CD62E (E-Selectin) |
| Cross Reactivity | Mouse |
| Includes | Mouse soluble CD62E (E-Selectin) standard, Mouse CD62E (E-Selectin) PE detection reagent, Mouse CD62E (E-Selectin) capture bead E6 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse CD62E (E-Selectin) flex set (bead E6) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction