Learn More
BD Cytometric Bead Array (CBA) Human TNFRII; Flex Set, Bead C5
Product Code. 15889968
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Product Code. 15889968
Brand: BD 560155
Includes
Human TNFRII; Standard; Human TNFRII; PE Detection Reagent; Human TNFRII; Capture Bead C5
Human reactivity was determined by testing samples with the BD CBA Human TNFRII Flex Set
  • Capable of measuring soluble human type 2 tumor necrosis factor receptor (TNFRII), also known as CD120b, in serum, plasma, and cell culture supernatant samples
TRUSTED_SUSTAINABILITY
Specifications
Antigen TNFRII
Cross Reactivity Human
Includes Human TNFRII, standard, Human TNFRII, PE detection reagent, Human TNFRII, capture bead C5
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human soluble TNFRII flex set (bead C5)