Learn More
BD Cytometric Bead Array (CBA) Human TNFRI ; Flex Set, Bead C4
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human TNFRI Standard; Human TNFRI PE Detection Reagent; Human TNFRI Capture Bead C4
Human reactivity was determined by testing samples with the BD™ CBA Human TNFRI Flex Set.
- Reactivity: QC Tested: Human
- Size: 100 tests
Specifications
Specifications
| Antigen | TNFRI |
| Cross Reactivity | Human |
| Includes | Human TNFRI standard, Human TNFRI PE detection reagent, Human TNFRI capture bead C4 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human soluble TNFRI flex set (bead C4) |
For Research use only.