Learn More
BD Cytometric Bead Array (CBA) Human TNF; Flex Set, Bead C4
Product Code. 15859938
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Product Code. 15859938
Brand: BD 560112
Includes
Human TNF; Standard; Human TNF; PE Detection Reagent; Human TNF; Capture Bead C4
Human and non-human primate reactivity was determined by testing samples with the BD CBA Human TNF Flex Set
  • Capable of measuring human tumor necrosis factor (TNF), formerly known as tumor necrosis factor-α, in serum, plasma, and cell culture supernatant samples
TRUSTED_SUSTAINABILITY
Specifications
Antigen TNF
Cross Reactivity Human
Includes Human TNF, standard, Human TNF, PE detection reagent, Human TNF, capture bead C4
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human TNF flex set (bead C4)