missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human TNF Enhanced Sensitivity Flex Set (Bead C4)

Product Code. 15801099
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

Product Code. 15801099

Brand: BD 561516

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

Includes

  • Kit contents:
  • Enhanced Sensitivity Capture Bead C4: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
  • Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
  • Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150

Measures human Tumor Necrosis Factor (TNF) in serum, plasma and cell culture supernatant samples

This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.

  • Bead-based immunoassay
  • Reactivity: human
  • Standard assay range: 274 to 200000fg/mL

Flow Cytometry/Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Cross Reactivity Human
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Human tumor necrosis factor, flow cytometry and immunoassay
Product Type Human TNF enhanced sensitivity flex set (bead C4)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.