Learn More
BD Cytometric Bead Array (CBA) Human MIG; Flex Set, Bead E8
Product Code. 15889278
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Product Code. 15889278
Brand: BD 558286
Includes
Human MIG; Standard; Human MIG; PE Detection Reagent; Human MIG; Capture Bead E8
Bead-based immunoassay capable of measuring human monokine-induced by interferon-γ (MIG), also known as CXCL9, in serum, plasma, and cell culture supernatant samples
Human reactivity was determined by testing samples with the BD CBA Human MIG Flex Set.
TRUSTED_SUSTAINABILITY
Specifications
Antigen MIG
Cross Reactivity Human
Includes Capture bead, detection reagent and standard
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
For Use With (Application) Flow Cytometry
Product Type Human MIG flex set (bead E8)
compliance-icons
Product Identifier
  • Human MIG Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT