Learn More
BD Cytometric Bead Array (CBA) Human G-CSF; Flex Set, Bead C8
Description
Includes
Human G-CSF; Standard; Human G-CSF; PE Detection Reagent; Human G-CSF; Capture Bead C8
The BD CBA Human G-CSF Flex Set is a bead-based immunoassay capable of measuring human Granulocyte-Colony Stimulating Factor (G-CSF) in serum, plasma, and cell culture supernatant samples. Human reactivity was determined by testing samples with the BD CBA Human G-CSF Flex Set.
- Reactivity: Human
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
Specifications
| Antigen | G-CSF |
| Cross Reactivity | Human |
| Includes | Human G-CSF, standard, Human G-CSF, PE detection reagent, Human G-CSF, capture bead C8 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human G-CSF flex set (bead C8) |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.