Learn More
BD Cytometric Bead Array (CBA) Human CD14; Flex Set, Bead D7
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human CD14; Standard; Human CD14; PE Detection Reagent; Human CD14; Capture Bead D7
The BD CBA Human CD14 Flex Set is a bead-based immunoassay capable of measuring human soluble CD14 in cell culture supernatant samples
- Reactivity: QC Testing: Human
- Bead Position: D7
- Limit of detection: 9.2pg/mL
With BD CBA human soluble protein master buffer kit (Cat. No. 558264, 100 tests, or 558265, 500 tests), flow cytometer, and FCAP Array™software
Specifications
Specifications
| Antigen | CD14 |
| Cross Reactivity | Human |
| Includes | Human CD14, standard, Human CD14, PE detection reagent, Human CD14, capture bead D7 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human soluble CD14 flex set (bead D7) |