missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human CD121b (IL-1 RII); Flex Set, Bead B7

Product Code. 15890069
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Product Code. Quantity unitSize
15890069 100 Tests Each
1 options
This item is not returnable. View return policy

Product Code. 15890069

Brand: BD 560281

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Human CD121b (IL-1 RII), Standard; PE Detection Reagent; Capture Bead B7.

For measuring soluble human soluble CD121b in serum, plasma, and cell culture supernatant samples

  • Also known as IL-1 Receptor Type-II
  • Bead-based immunoassay
  • Human reactivity determined by testing samples with BD CBA Human Soluble CD121b Flex Set

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD121beta (IL-1 RII)
Cross Reactivity Human
Includes Human CD121b (IL-1 RII), standard, PE detection reagent, capture bead B7
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human CD121b (IL-1 RII) flex set (bead B7)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.