missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD CBA Mouse Immunoglobulin Isotyping Kit
Description
Includes
Heavy and light chain isotypes of mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM, and IgE; reagents for capture of mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM, and IgE; anti-kappa and anti-lambda detection reagents; standards; buffers; cytometer setup reagents
- Provides a capture surface for a specific protein and is analogous to an individually coated well in an ELISA plate
- Broad dynamic range of fluorescence detection, combined with multiple analyte detection, allows for small sample volume, fewer sample dilutions, and substantially less time overall to establish the value of an unknown versus a conventional ELISA approach
- Can be used to rapidly identify the heavy and light chain isotypes of a mouse monoclonal antibody in a single sample
- Contains enough reagents to perform 100 tests
Specifications
Specifications
| Antigen | IgG |
| Cross Reactivity | Mouse |
| Includes | Heavy and light chain isotypes of Mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM and IgE; reagents for capture of Mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM and IgE, Anti-kappa and Anti-lambda detection reagents, standards, Buffers, cytometer setup reagents |
| Quantity | 100 Tests |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse Immunoglobulin isotyping kit |
For research use only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?