Learn More
BD Caspase-3 Enzyme Set
Product Code. 15898708
Change view
Click to view available options
Cross Reactivity:
Human
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cross Reactivity Unit Size
15898708 Human Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15898708 Supplier BD Supplier No. 556473
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Recombinant Human Active Caspase-3: 5μg, Ac-DEVD-AFC, Caspase-3 Fluorogenic Substrate: 1.0mg, Ac-DEVD-CHO, Caspase-3 Inhibitor: 1.0mg
Gene that is required for normal apoptosis during development
Caspases are cysteine proteases that play central role in apoptosis. The caspase family was discovered by searching human cDNA libraries for sequences homologous to ced-3, a C. The rate of caspase-3 enzymatic hydrolysis can be measured by release of AFC, from Ac-DEVD-AFC caspase substrate, through emission at 480-520nm (peak at 505nm) upon excitation at 400nm using UV spectrofluorometry
  • Consisting of 12kDa and 17kDa subunit derived from 32kDa pro-enzyme (pro- caspase-3)
  • Cleaves PARP, an enzyme that is involved in DNA repair and genomic maintenance
  • The cleavage site in PARP is C-terminal to Asp-216
  • Cleaves the tetrapeptide between D and AFC, thus releasing the fluorogenic AFC
  • Expressed in E.coli and purified
Enzyme assay
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Enzyme Assay
Name Caspase-3 Enzyme Set
Quantity 20 Tests
Storage Requirements -80°C
Cross Reactivity Human