Learn More
BD BBL™ AFB Slides
Product Code. 11722964
Change view
Click to view available options
Quantity:
50 Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11722964 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11722964 Supplier BD Supplier No. 231391
 Restricted Item
Estimated Shipment: 01-12-2026
For evaluating and controlling the quality of acid-fast and fluorescent stain reagents and staining techniques for the detection of acid-fast bacilli (AFB)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Control Slide
Quantity 50 Pk.
For in vitro diagnostic use.