Learn More
BD Bacto™ Dehydrated Agar
Product Code. 11768223
Change view
Click to view available options
Quantity:
100 g
454 g
2000 g
Packaging Type:
Bottle
Container
Unit Size:
100g
2kg
454g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11768223 100 g Bottle 100g
10455513 454 g Bottle 454g
11758223 2000 g Container 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11768223 Supplier BD Supplier No. 214050
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
  • Extraneous matter, pigmented portions and salts are reduced to minimum
  • Fine, light colored granules
  • Convenient for weighing and handling
Determines motility and growth of anaerobes and microaerophiles
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO/DIS, 11014
Concentration 1.5%
Additive Agar
Form Dehydrated
For Use With (Application) Determination of motility and the growth of anaerobes and microaerophiles
Packaging Type Bottle
Quantity 100 g