Beschreibung
Beinhaltet
Kit Part A:
- PerCP-Cy5.5 Mouse Anti-BrdU
- Alexa Fluor™ 647 Mouse Anti-H2AX (pS139)
- PE Mouse Anti-Cleaved PARP (Asp214) Antibody
- Cytofix/Cytoperm Fixation/Permeabilization Solution
- Perm/Wash Buffer (10X)
- Cytofix/Cytoperm Plus Permeabilization Buffer
- DAPI
- BrdU (10mg/mL)
- DNase
Multiparameter flow cytometry provides a powerful tool for resolving mechanisms by which individual cells in homogenous or mixed cell populations maintain viability, enter and progress through cell cycle or undergo cell death. These probes along with optimized protocols enable multicolor flow cytometric analysis of proliferation, DNA damage and apoptosis, respectively, by individual cells within samples.
- Consists of Apoptosis, DNA Damage and Cell Proliferation Kit - Part A and Apoptosis, DNA Damage and Cell Proliferation Kit - Part B
Spezifikation
Spezifikation
| Content And Storage | Kit Part A: 4°C and Kit Part B: -80°C |
| For Use With (Application) | Flow Cytometry, Cell Proliferation |
| Quantity | 50 Tests |
| Reactivity | Human |