missing translation for 'onlineSavingsMsg'
Learn More

Bazedoxifene, TRC

Product Code. 30136361
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136361 50 mg 50mg
17907854 5 mg 5mg
17949364 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136361 Supplier Toronto Research Chemicals Supplier No. TRCB12924550MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198481-32-2
Molecular Formula C30 H34 N2 O3
Molecular Weight (g/mol) 470.6
Synonym 1H-Indol-5-ol, 1-[[4-[2-(hexahydro-1H-azepin-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-, 1-[[4-[2-(Hexahydro-1H-azepin-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-1H-indol-5-ol, Bazedoxifene, WAY 140424
IUPAC Name 1-[[4-[2-(azepan-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methylindol-5-ol
SMILES OC=1C=CC(=CC1)C2=C(C3=CC(O)=CC=C3N2CC4=CC=C(OCCN5CCCCCC5)C=C4)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C30H34N2O3/c1-22-28-20-26(34)12-15-29(28)32(30(22)24-8-10-25(33)11-9-24)21-23-6-13-27(14-7-23)35-19-18-31-16-4-2-3-5-17-31/h6-15,20,33-34H,2-5,16-19,21H2,1H3
Formula Weight 470.2569
Chemical Name or Material Bazedoxifene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.