Learn More
Bazedoxifene-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Bazedoxifene-d4 Acetate |
| CAS | 1133695-49-4 |
| Molecular Formula | C30 H30 D4 N2 O3 |
| Quantity | 10 mg |
| Synonym | Bazedoxifene-d4 Acetate, 1-[[4-[2-(Hexahydro-1H-azepin-1-yl)(ethoxy-d4)]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-1H-indol-5-ol Acetate, TSE 424-d4, Viviant-d4, WAY-TES 424-d4 |
| InChI Formula | InChI=1S/C30H34N2O3/c1-22-28-20-26(34)12-15-29(28)32(30(22)24-8-10-25(33)11-9-24)21-23-6-13-27(14-7-23)35-19-18-31-16-4-2-3-5-17-31/h6-15,20,33-34H,2-5,16-19,21H2,1H3/i18D2,19D2 |
| SMILES | [2H]C([2H])(Oc1ccc(Cn2c(c(C)c3cc(O)ccc23)c4ccc(O)cc4)cc1)C([2H])([2H])N5CCCCCC5 |
| IUPAC Name | 1-[[4-[2-(azepan-1-yl)-1,1,2,2-tetradeuterioethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methylindol-5-ol |
| Molecular Weight (g/mol) | 474.627 |
| Formula Weight | 474.282 |