missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BAY 826
GREENER_CHOICE

Product Code. 18786316
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18786316

Brand: Tocris Bioscience™ 6579/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent Tie2 inhibitor

BAY 826 is a potent Tie 2 inhibitor (IC50 = 1.3 nM for autophosphorylation of Tie2 in HUVECs), which is selective for Tie2 over other angiogenic receptor tyrosine kinases, including VEGFR, FGFR and PDGFR. BAY 826 displays in vivo efficacy in some murine glioma models.

Tie2 is also known as angiopoietin-1 receptor and TEK tyrosine kinase.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [3-Cyano-5-[[[2,4-dimethyl-5-[6-(3-pyridinyl)-1H-imidazo[1,2-b]pyrazol-1-yl]phenyl]amino]carbonyl]phenyl]pentafluorosulfur
CAS 1448316-08-2
Quantity 5 mg
Molecular Formula C26H19F5N6OS
Formula Weight 558.53
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.