missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BAY 826

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
BAY 826 is a potent Tie 2 inhibitor (IC50 = 1.3 nM for autophosphorylation of Tie2 in HUVECs), which is selective for Tie2 over other angiogenic receptor tyrosine kinases, including VEGFR, FGFR and PDGFR. BAY 826 displays in vivo efficacy in some murine glioma models.
Tie2 is also known as angiopoietin-1 receptor and TEK tyrosine kinase.
Specifications
Specifications
| Chemical Name or Material | [3-Cyano-5-[[[2,4-dimethyl-5-[6-(3-pyridinyl)-1H-imidazo[1,2-b]pyrazol-1-yl]phenyl]amino]carbonyl]phenyl]pentafluorosulfur |
| CAS | 1448316-08-2 |
| Quantity | 25 mg |
| Molecular Formula | C26H19F5N6OS |
| Formula Weight | 558.53 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction