Learn More
Lovibond™ Battery
Product Code. 10039951
Change view
Click to view available options
Battery Type:
9 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Battery Type Unit Size
10039951 9 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10039951 Supplier Lovibond™ Supplier No. 1950012
  • ONLINE EXCLUSIVE PRICE
    3.80 EUR
    3.70 EUR / Pack of 1
    Save 0.10 EUR
    2% Off
Estimated Shipment: 14-12-2026
Order Info
BatteryType
Specifications
Battery Type 9 V
For Use With (Equipment) Photometer
Quantity 1 Ea.
Voltage 9 V