missing translation for 'onlineSavingsMsg'
Learn More

BAT3/BAG6 Antibody [CoraFluor™ 1], Novus Biologicals™

Product Code. 30753752 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30753752 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30753752 Supplier Novus Biologicals Supplier No. AF6438CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

BAT3/BAG6 Polyclonal antibody specifically detects BAT3/BAG6 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAT3/BAG6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias BAT3HLA-B associated transcript-3, BCL2-associated athanogene 6BAG-6, D6S52EHLA-B-associated transcript 3, G3BAG family molecular chaperone regulator 6, HLA-B associated transcript 3, large proline-rich protein BAT3, Protein G3, Protein Scythe, scythe
Host Species Sheep
Immunogen E. coli-derived recombinant human BAT3/BAG6, Met1-Gln190, Accession # P46379
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 7917
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.