Learn More
MP Biomedicals™ Basal Medium Eagle (Modified), with Hanks' Salts and L-Glutamine, without Sodium Carbonate
Shop All MP Biomedicals ProductsDescription
- Basal Medium Eagle (BME) is one of the most commonly used of all cell culture media.
- This medium is the result of numerous studies from the late 1950's analyzing the nutrient requirements and other components essential for the growth of cells in culture.
- Traditionally, BME has been used in studies to measure the growth response of normal (WI-38) and transformed (HeLa) cells in monolayer culture.
- BME may be used for a broad variety of cell lines when properly supplemented.
- It has a pH of 6.5 ± 0.5 (w/o sodium bicarb.), 7.3 ± 0.5 (adjusted with sodium bicarb.)
Specifications
Specifications
| Form | Powder |
| For Use With (Application) | Bacteria Cell Culture, Mammalian Cell Culture |
| Quantity | 10 L |
| Formulation | Polypeptone 10.0, Biotryptase 10.0, Heart Peptone 3.0, Corn Starch 1.0, Sodium Chloride 5.0, Agar 13.5, Final pH 7.3 ±0.2 |
| pH | 6.5 to 7.3 ±0.5 |
| Preparation Method | Suspend 42.5 g of the medium in one liter of distilled or deionized water. Mix well and heat with frequent agitation. Boil for one minute. Dispense and sterilize in an autoclave at 121°C (15 lb. psi) for 15 minutes. Cool to 45 to 50°C and add 5 to 10% sterile defibrinated sheep or rabbit blood if desired. |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?