Learn More
Buerkle™ Barrel Valve
Product Code. 15655494 Shop All Bürkle Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15655494 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15655494 Supplier Buerkle™ Supplier No. 05360010
  • ONLINE EXCLUSIVE PRICE
    139.00 EUR
    132.65 EUR / Pack of 1
    Save 6.35 EUR
    4% Off
Estimated Shipment: 18-12-2026
Enables quick and clean emptying even with highly viscous liquids, suitable for metal barrel with 2 in. BSP TriSure metal barrel screw joint
  • Glass fibre reinforced PP
  • Seals NBR
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
For Use With (Equipment) Barrel with G 2 in.
Material Polypropylene
Product Type Spigot