Learn More
Gilac™ Barrel with Tab Lid
Product Code. 17958561
Change view
Click to view available options
Capacity (Metric):
50 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17958561 50 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17958561 Supplier Gilac™ Supplier No. G032208
  • ONLINE EXCLUSIVE PRICE
    69.67 EUR
    50.45 EUR / Pack of 1
    Save 19.22 EUR
    27% Off
Estimated Shipment: 20-11-2026
Barrel with Tab Lid is a food contact certified plastic barrel, suitable for a large number of uses (brining, pickling etc.) when transporting and storing liquid or solid food products.
  • Food contact certified plastic barrel.
  • Suitable for a large number of uses (brining, pickling etc.) when transporting and storing liquid or solid food products.
  • Global and specific migration test approved.
  • Contents are protected from contamination thanks to its watertight lid.
  • Handling is easy and can be opened completely.
Specifications
Product Type Barrel
Capacity (English) 13.2 gal.
Capacity (Metric) 50 L
Color White
For Use With (Application) Alimentary or dangerous liquid
Material HDPE