Learn More
Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps
Product Code. 11367131
Change view
Click to view available options
For Use With (Equipment):
GenPure (UV, UV/UF, UV-TOC and UF/UV-TOC Only), LabTower EDI
LabTower RO
MicroPure, Smart2Pure (UV and UV/UVF Only)
Pacific TII; LabTower TII
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11367131 GenPure (UV, UV/UF, UV-TOC and UF/UV-TOC Only), LabTower EDI Each
11357111 MicroPure, Smart2Pure (UV and UV/UVF Only) Each
11367171 LabTower RO Each
16476390 Pacific TII; LabTower TII Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11367131 Supplier Thermo Scientific™ Supplier No. 09.2002
Estimated Shipment: 26-01-2027
Maintain UV function with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps.
Maintain UV function with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps, for use with Barnstead point of use water purification systems.
Ideal for use with the following Barnstead point of use water purification systems:
09.1002:
  • MicroPure™ (UV and UV/UVF systems only)
  • Smart2Pure™ (UV and UV/UVF systems only)
09.2002:
  • GenPure™ (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
  • GenPure™ Pro (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
  • GenPure™ xCAD (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
  • LabTower™ EDI
09.4002:
  • Pacific™ TII
  • LabTower™ TII
09.5002:
For storage reservoirs (230V only) used with the following Barnstead water purification systems:
  • LabTower RO
  • LabTower TII
  • Pacific TII (30 L, 60 L, 100 L only)
  • Smart2Pure (12 only)
TRUSTED_SUSTAINABILITY
Specifications
Description Replacement UV lamp
For Use With (Equipment) GenPure (UV, UV/UF, UV-TOC and UF/UV-TOC Only), LabTower EDI
Type UV Lamp