Learn More
Barium Titanate (IV), Sigma-Aldrich™
Specifications
Specifications
| Melting Point | 1625°C |
| CAS | 12047-27-7 |
| Quantity | 250 g |
| Density | 6.08 g/cm3 at 20°C |
| MDL Number | MFCD00003447 |
| Synonym | Barium titanate (IV), Barium metatitanate, Barium titanate (IV) |
| Molecular Weight (g/mol) | 233.19 |
| Grade | Research |
| Chemical Name or Material | Barium Titanate (IV) |