Learn More
Barium Sulfate, Multi-Compendial, U.S.P., J.T. Baker™
Product Code. 15587884 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15587884 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15587884 Supplier Avantor J.T.Baker Supplier No. 104101
Specifications
CAS 7727-43-7
Quantity 500 g
Molecular Formula BaO4S
MDL Number MFCD00003455
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
SMILES [Ba++].[O-]S([O-])(=O)=O
IUPAC Name barium(2+);sulfate
Molecular Weight (g/mol) 233.38
PubChem CID 24414
Formula Weight 233.39
Grade Multi-Compendial/USP
Packaging Poly Bottle
Specific Gravity 1L = 4.50kg
Chemical Name or Material Barium Sulfate