Learn More
Barium Hexafluorosilicate, TRC
Specifications
Specifications
| Chemical Name or Material | Barium Hexafluorosilicate |
| CAS | 17125-80-3 |
| Molecular Formula | BaSiF6 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/Ba.F6Si/c;1-7(2,3,4,5)6/q+2;-2 |
| SMILES | [Ba+2].F[Si-2](F)(F)(F)(F)F |
| IUPAC Name | barium(2+);hexafluorosilicon(2-) |
| Molecular Weight (g/mol) | 279.4 |
| Formula Weight | 279.8726 |