Learn More
Barium Chloride Dihydrate, Supelco™
Product Code. 15884110
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884110 5 kg 5kg
15864110 500 g 500g
15874110 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15884110 Supplier Supelco Supplier No. 1017195000
 Restricted Item
Estimated Shipment: 25-11-2026
For analysis EMSURE™ ACS,ISO,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10326-27-9
Grade Research
Molecular Weight (g/mol) 244.26
Specifications
Quantity 5 kg
MDL Number MFCD00149154
Synonym Barium chloride dihydrate, Barium dichloride, Barium chloride dihydrate
Density 3.86 g/cm3 at 20°C
Chemical Name or Material Barium Chloride Dihydrate