Learn More
Barium Acetate, Supelco™
Specifications
Specifications
| Melting Point | 450°C |
| CAS | 543-80-6 |
| Quantity | 500 g |
| Molecular Formula | Ba(CH3COO)2 |
| MDL Number | MFCD00012447 |
| Synonym | Barium acetate, Barium acetate |
| Molecular Weight (g/mol) | 255.42 |
| Grade | Research |
| Density | 2.47 g/cm3 at 20°C |
| Chemical Name or Material | Barium Acetate |