Learn More
Cole-Parmer™ Balance Draft Shield
Product Code. 12891763 Shop All Cole Parmer Products
Change view
Click to view available options
Width (Metric):
330 mm
381 mm
457 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
12891763 381 mm Each
15380645 330 mm Each
15300655 457 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12891763 Supplier Cole-Parmer™ Supplier No. 0101904
  • ONLINE EXCLUSIVE PRICE
    230.00 EUR
    202.74 EUR / Pack of 1
    Save 27.26 EUR
    11% Off
View all promotions
Estimated Shipment: 25-01-2027
Clear acrylic prevents drafts from affecting weighings
  • No assembly required
  • Mouse hole in back panel for power cords
  • Raised edges on top of enclosure allow secure placement of frequently used weighing accessories
  • Minimizes air movement around your balance for faster, more stable readings
Specifications
Color Clear
Width (Metric) 381 mm