missing translation for 'onlineSavingsMsg'
Learn More

Bafilomycin A1, MedChemExpress

Product Code. 16464376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464376 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16464376 Supplier Medchem Express Supplier No. HY100558
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Bafilomycin A1 (BafA1) is a specific and reversible inhibitor of vacuolar H+-ATPase (V-ATPase) with IC50 values of 4-400 nmol/mg. Bafilomycin A1, a macrolide antibiotic, is also used as an autophagy inhibitor at the late stage. Bafilomycin A1 blocks autophagosome-lysosome fusion and inhibits acidification and protein degradation in lysosomes of cultured cells. Bafilomycin A1 induces apoptosis.

Bafilomycin A1 (BafA1) is a specific and reversible inhibitor of vacuolar H+-ATPase (V-ATPase) with IC50 values of 4-400 nmol/mg. Bafilomycin A1, a macrolide antibiotic, is also used as an autophagy inhibitor at the late stage. Bafilomycin A1 blocks autophagosome-lysosome fusion and inhibits acidification and protein degradation in lysosomes of cultured cells. Bafilomycin A1 induces apoptosis.

Specifications

Chemical Name or Material Bafilomycin A1
Physical Form Powder
CAS 88899-55-2
Color White
Quantity 100 μg
Purity Grade Notes Research
Molecular Formula C35H58O9
For Use With (Application) Cancer-programmed cell death
Synonym BafA1
Solubility Information DMSO : 100 mg/mL (160.56 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES C[C@H]([C@](O[C@@H]1C(C)C)(C[C@@H](O)[C@@H]1C)O)[C@H](O)[C@@H]([C@](OC(/C(OC)=C/C(C)=C\[C@@H](C)[C@@H](O)[C@@H](C)C2)=O)([H])[C@H](/C=C/C=C2\C)OC)C
Molecular Weight (g/mol) 622.83
Formula Weight 622.83
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.