Learn More
Brady™ B33 Series Self-laminating White Polyester Labels
Description
For BBP33/i3300 printers.
Specifications
Specifications
| Adhesive Type | Permanent Acrylic |
| Color | White |
| Finish | Matte |
| Format | Roll |
| Material | Polyester |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| Sterility | Non-sterile |
| Dimensions (L x W) | 15.24 mm (Dia) |
| Length (English) | 15.24 mm |
| Show More |