Learn More
Lonza™ b-G-Blocker
Product Code. 11660301
Change view
Click to view available options
Quantity:
5 x 5 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11660301 5 x 5 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11660301 Supplier Lonza™ Supplier No. N190
  • ONLINE EXCLUSIVE PRICE
    590.00 EUR
    499.84 EUR / 1 sets
    Save 90.16 EUR
    15% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Glucan blocker to prevent false positive results in LAL assays.
  • Glucan blocker to prevent false positive results during LAL assays.
  • Some examples of glucan sources include yeast and cellulosic materials including hemodialysis filters.
  • Product may be used with any of our LAL assays. Note: PYROGENT™ Gel Clot LAL products typically do not require ß-G-Blocker.
TRUSTED_SUSTAINABILITY
Specifications
Assays LAL assays
Format Vials
For Use With (Application) Blocking glucan for LAL assays
Quantity 5 x 5 mL