Learn More
Aztreonam Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Aztreonam Ethyl Ester |
| CAS | 102579-57-7 |
| Molecular Formula | C15 H21 N5 O8 S2 |
| Quantity | 5 mg |
| Synonym | Aztreonam Ethyl Ester |
| InChI Formula | InChI=1S/C15H21N5O8S2/c1-5-27-13(23)15(3,4)28-19-10(8-6-29-14(16)17-8)11(21)18-9-7(2)20(12(9)22)30(24,25)26/h6-7,9H,5H2,1-4H3,(H2,16,17)(H,18,21)(H,24,25,26)/b19-10-/t7-,9-/m0/s1 |
| SMILES | CCOC(=O)C(C)(C)O\N=C(/C(=O)N[C@H]1[C@H](C)N(C1=O)S(=O)(=O)O)\c2csc(N)n2 |
| IUPAC Name | (2S,3S)-3-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(1-ethoxy-2-methyl-1-oxopropan-2-yl)oxyiminoacetyl]amino]-2-methyl-4-oxoazetidine-1-sulfonic acid |
| Molecular Weight (g/mol) | 463.486 |
| Formula Weight | 463.083 |