Learn More
Azlon™ Polycarbonate Vacuum Desiccator with Stopcock
Product Code. 12060253
Change view
Click to view available options
Height (Metric):
262 mm
296 mm
Diameter (Metric) Plate:
205 mm
236 mm
Diameter (Metric) Inner:
215 mm
245 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Diameter (Metric) Plate Diameter (Metric) Inner unitSize
12060253 262 mm 205 mm 215 mm Each
12080253 296 mm 236 mm 245 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12060253 Supplier Azlon Supplier No. DWA215
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
Durable with high impact resistance and safely withstands high vacuum pressures
Durable with high impact resistance and safely withstands high vacuum pressures
  • Transparent polycarbonate cover and base
  • Durable with high impact resistance
  • Will safely withstand high vacuum pressures
  • Ceramic plate permits the use of incandescent crucibles
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Material Polycarbonate
Product Type Vacuum Dessicator
Includes Stopcock
Quantity 1/Pk.
Height (Metric) 262 mm
Height (Metric) Interior 178 mm
Diameter (Metric) Plate 205 mm
Diameter (Metric) Inner 215 mm